Skip to Content
Merck
All Photos(3)

Key Documents

250309

Sigma-Aldrich

4-Amino-1,1′-azobenzene-3,4′-disulfonic acid monosodium salt

Dye content 90 %

Synonym(s):

2-Amino-5-([4-sulfophenyl]azo)benzenesulfonic acid monosodium salt, Acid Yellow 9 monosodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
RO3SC6H4N=NC6H3(NH2)SO3R, R = H or Na
CAS Number:
Molecular Weight:
379.34
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

form

powder

composition

Dye content, 90%

λmax

489 nm

ε (extinction coefficient)

11438 at 489 nm in 0.1 M HCl at 0.01 g/L
15302 at 385 nm in 1 M HCl at 0.01 g/L

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].[Na+].Nc1ccc(cc1S(O)(=O)=O)\N=N\c2ccc(cc2)S([O-])(=O)=O.Nc3ccc(cc3S([O-])(=O)=O)\N=N\c4ccc(cc4)S(O)(=O)=O

InChI

1S/2C12H11N3O6S2.2Na/c2*13-11-6-3-9(7-12(11)23(19,20)21)15-14-8-1-4-10(5-2-8)22(16,17)18;;/h2*1-7H,13H2,(H,16,17,18)(H,19,20,21);;/q;;2*+1/p-2/b2*15-14+;;

InChI key

MPOAAHDXCIUENZ-XNDFEMONSA-L

General description

4-Amino-1,1′-azobenzene-3,4′-disulfonic acid monosodium salt, also called as acid yellow 9, belongs to the class of azo dyes.[1]

Application

4-Amino-1,1′-azobenzene-3,4′-disulfonic acid monosodium salt has been used as a control to study the influence of aromatic substitution patterns on azo dye degradability by Streptomyces spp and the fungus Phanerochaete chrysosporium.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Influence of Aromatic Substitution Patterns on Azo Dye Degradability by Streptomyces spp. and Phanerochaete chrysosporium
GRIGSBY M B P
Applied and Environmental Microbiology, 58, 3605-3613 (1992)
J Cuamatzi-Flores et al.
PloS one, 14(1), e0209711-e0209711 (2019-01-05)
Dye-decolorizing peroxidase (DyP) from the white rot basidiomycete Pleurotus ostreatus is a heme peroxidase able to oxidize diverse substrates, including recalcitrant phenols and dyes. This study analyzed the effect of chemical dyes on P. ostreatus growth, DyP activity and the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service