Skip to Content
Merck
All Photos(2)

Key Documents

248959

Sigma-Aldrich

Bis(2-ethylhexyl) phosphite

96%

Synonym(s):

Bis(2-ethylhexyl) hydrogen phosphite, Bis(2-ethylhexyl) phosphonate, Bis(2-ethylhexyl) phosphonic acid, Di-2-Ethylhexyl phosphite, Di-2-ethylhexyl hydrogen phosphite, Diisooctyl phosphite, Phosphonic acid di(2-ethylhexyl) ester

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
R$1,048.00

R$1,048.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
R$1,048.00

About This Item

Linear Formula:
[CH3(CH2)3CH(C2H5)CH2O]2P(O)H
CAS Number:
Molecular Weight:
306.42
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

R$1,048.00


Check Cart for Availability

Request a Bulk Order

Assay

96%

form

liquid

refractive index

n20/D 1.442 (lit.)

density

0.916 g/mL at 25 °C (lit.)

SMILES string

CCCCC(CC)CO[PH](=O)OCC(CC)CCCC

InChI

1S/C16H35O3P/c1-5-9-11-15(7-3)13-18-20(17)19-14-16(8-4)12-10-6-2/h15-16,20H,5-14H2,1-4H3

InChI key

HZIUHEQKVCPTAJ-UHFFFAOYSA-N

General description

Bis(2-ethylhexyl) phosphite is an chemical warfare simulant and its reaction with common ion binary ionic liquid has been investigated[1]. Mass-transfer kinetics of Eu3+ and Am3+ in bis(2-ethylhexyl) phosphite-n-dodecane-NaCl-HCl-water system has been investigated[2].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reactions of Chemical Warfare Agent Simulants in the Common Ion Binary Ionic Liquid 1, 2-Dimethyl-3-propylimidazolium/Copper (II) Bis (trifluoromethylsulfonyl) amide.
Wilkes JS, et al.
Industrial & Engineering Chemistry Research, 48(13), 6203-6211 (2009)
Kinetics and mechanism of the interfacial mass transfer of europium (3+) and americium (3+) in the system bis (2-ethylhexyl) phosphate-n-dodecane-sodium chloride-hydrochloric acid-water.
Danesi PR and Vandegrift GF.
The Journal of Physical Chemistry, 85(24), 3646-3651 (1981)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service