Skip to Content
Merck
All Photos(1)

Key Documents

246921

Sigma-Aldrich

1,8-Bis(bromomethyl)naphthalene

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H6(CH2Br)2
CAS Number:
Molecular Weight:
314.02
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

form

crystals

mp

131-133 °C (lit.)

functional group

bromo

SMILES string

BrCc1cccc2cccc(CBr)c12

InChI

1S/C12H10Br2/c13-7-10-5-1-3-9-4-2-6-11(8-14)12(9)10/h1-6H,7-8H2

InChI key

GCZOMCDXYFMAGP-UHFFFAOYSA-N

General description

Two-photon chemistry of 1,8-bis(bromomethyl)naphthalene has been studied by time-delayed, two-color photolysis and argon ion laser-jet photolysis techniques[1].

Application

1,8-Bis(bromomethyl)naphthalene has been used in the synthesis of indan-based unusual α-amino acid derivatives under solid-liquid phase-transfer catalysis conditions[2].

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Two-Photon Laser-Induced Reaction of 1, 8-Bis (halomethyl) naphthalenes from Different Excited States and Transient Targeting of Its Intermediate by Time-Delayed, Two-Color Photolysis and Argon Ion Laser-Jet Photolysis Techniques
Ouchi A, et al.
Journal of the American Chemical Society, 119(3), 592-599 (1997)
S Kotha et al.
The Journal of organic chemistry, 65(5), 1359-1365 (2000-05-18)
Conformationally constrained cyclic alpha-amino acid derivatives were synthesized under solid-liquid phase-transfer catalysis conditions. This methodology involves the bis-alkylation of ethyl isocyanoacetate with various alpha,alpha'-dibromo-o-xylene derivatives [alpha,alpha'-dibromo-o-xylene 5, 2,3-bis(bromomethyl)-1, 4-dimethoxybenzene 6, 1,2-bis(bromomethyl)-4,5-dibromobenzene 7, 2, 3-bis(bromomethyl)naphthalene 8, 1,8-bis(bromomethyl)-naphthalene 9, 6,7-bis(bromomethyl)-2,2-dimethyl-1H-phenalene-1,3(2H)-dione 10, 2

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service