Skip to Content
Merck
All Photos(1)

Key Documents

246263

Sigma-Aldrich

Indole-4-carboxylic acid

98%

Synonym(s):

4-Carboxyindole

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
R$1,644.00

R$1,644.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
R$1,644.00

About This Item

Empirical Formula (Hill Notation):
C9H7NO2
CAS Number:
Molecular Weight:
161.16
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

R$1,644.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

powder

mp

213-214 °C (lit.)

SMILES string

OC(=O)c1cccc2[nH]ccc12

InChI

1S/C9H7NO2/c11-9(12)7-2-1-3-8-6(7)4-5-10-8/h1-5,10H,(H,11,12)

InChI key

ROGHUJUFCRFUSO-UHFFFAOYSA-N

Application

tert-butyl esters of indole and pyrrolecarboylic acids are readily prepared by direct treatment with N,N-dimethylformamide di-tert-butyl acetal (Cat. No. 395005).[1]
  • Reactant for preparation of substituted indole derivatives as histamine H3 antagonists
  • Reactant for preparation of potent and selective inhibitors of human reticulocyte 15-lipoxygenase-1
  • Reactant for preparation of inhibitors of Gli1-mediated transcription in Hedgehog pathway
  • Reactant for preparation of pyridinyl carboxylates as SARS-CoV 3CL proinhibitors
  • Reactant for preparation of substituted bipiperidinylmethyl amides as CCR3 membrane binding ligands
  • Reactant for preparation of indole amide hydroxamic acids as potent inhibitors of histone deacetylases
  • Reactant for preparation of 2-[[[4′-chloro-[1,1-biphenyl]-4-yl]thio]methyl]-N-hydroxybutanamide derivatives as specific metalloproteinase

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

SOLVENT EFFECTS ON THE ELECTRONIC ABSORPTION AND FLUORESCENCE SPECTRA OF INDOLE?4?CARBOXYLIC ACID: PROTOTROPIC EQUILIBRIA IN AQUEOUS SOLUTIONS.
Krishnamurthy M.
Photochemistry and Photobiology, 45(3), 359-364 (1987)
Synthetic Communications, 34, 3691-3691 (2005)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service