Skip to Content
Merck
All Photos(2)

Key Documents

244775

Sigma-Aldrich

Triphenyltin hydride

Synonym(s):

Triphenylstannane, Triphenylstannyl hydride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)3SnH
CAS Number:
Molecular Weight:
351.03
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

form

solid

reaction suitability

reagent type: reductant

refractive index

n20/D 1.632 (lit.)

bp

163-165 °C/0.3 mmHg (lit.)

mp

28 °C (lit.)

density

1.374 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

c1ccc(cc1)[SnH](c2ccccc2)c3ccccc3

InChI

1S/3C6H5.Sn.H/c3*1-2-4-6-5-3-1;;/h3*1-5H;;

InChI key

NFHRNKANAAGQOH-UHFFFAOYSA-N

Application

Triphenyltin hydride can be used as a reducing agent in:
  • The reductive deselenenylation reaction in the presence of the catalytic amount of AIBN.
  • The free radical hydrostannylations of olefins.
  • The reduction of α, β-unsaturated ketones and esters.

It can also be used as a radical precursor in the free-radical reduction reactions in the presence of 9-borabicyclo[3.3.1]nonane (9-BBN) as an initiator.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jun Xu et al.
Ecotoxicology (London, England), 20(1), 73-80 (2010-10-29)
The toxicity of organotin compounds in the environment is closely related to their uptake by microorganisms and delivery through the food chain. The population at low trophic levels like microalgae plays an important role in this aspect. In this study
Li-Lian Liu et al.
Marine pollution bulletin, 63(5-12), 535-540 (2011-03-08)
The present study was undertaken to evaluate the distribution and accumulation of tributyltin (TBT) and triphenyltin (TPhT) in seawater, sediments and selected organisms from a cage mariculture area in southern Taiwan, Hsiao Liouchiou Island. Our results show that ΣOTs were
On the use of 9-borabicyclo [3.3. 1] nonane as an initiator for low-temperature free-radical reductions
Perchyonok VT and Schiesser CH
Tetrahedron Letters, 39(30), 5437-5438 (1998)
Triethylborane induced selective 1, 4-reduction of α, β-unsaturated carbonyl compounds with triphenyltin hydride or tributyltin hydride
Nozaki K, et al.
Bulletin of the Chemical Society of Japan, 64(8), 2585-2587 (1991)
Lin Yu et al.
Journal of hazardous materials, 192(3), 1860-1868 (2011-08-09)
Triphenyltin (TPT) has high binding affinity with the retinoid X receptor (RXR) in animals. The natural ligand of RXR, 9-cis-retinoic acid (RA), is known to induce featured malformations in vertebrate embryos by disrupting RA signal. Limited information is available on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service