Skip to Content
Merck
All Photos(3)

Key Documents

242446

Sigma-Aldrich

Suberic acid monomethyl ester

97%

Synonym(s):

Monomethyl suberate

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
R$422.00
5 G
R$1,771.00

R$422.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
R$422.00
5 G
R$1,771.00

About This Item

Linear Formula:
HO2C(CH2)6CO2CH3
CAS Number:
Molecular Weight:
188.22
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

R$422.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

refractive index

n20/D 1.444 (lit.)

bp

185-186 °C/18 mmHg (lit.)

mp

17-19 °C (lit.)

density

1.047 g/mL at 25 °C (lit.)

functional group

carboxylic acid
ester

SMILES string

COC(=O)CCCCCCC(O)=O

InChI

1S/C9H16O4/c1-13-9(12)7-5-3-2-4-6-8(10)11/h2-7H2,1H3,(H,10,11)

InChI key

KOVPXZDUVJGGFU-UHFFFAOYSA-N

General description

The effects of suberic acid monomethyl ester (monomethyl suberate) on liver lactate metabolism was studied[1].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Rognstad
Biomedica biochimica acta, 50(7), 921-930 (1991-01-01)
We have examined the effects of salicylate on fluxes of lactate metabolism in rat hepatocytes using a steady state model. Salicylate produces an uncoupling effect, an inhibition of gluconeogenesis, a marked activation of pyruvate dehydrogenase flux, and an inhibition of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service