Recommended Products
Assay
95%
form
liquid
refractive index
n20/D 1.481 (lit.)
bp
109-110 °C/2 mmHg (lit.)
density
1.337 g/mL at 25 °C (lit.)
SMILES string
ClCCCCCCBr
InChI
1S/C6H12BrCl/c7-5-3-1-2-4-6-8/h1-6H2
InChI key
JTYUIAOHIYZBPB-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
1-Bromo-6-chlorohexane was used in the synthesis of zinc reagent by reacting with Mg turnings in the presence of ZnCl2 and LiCl.
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
213.8 °F - closed cup
Flash Point(C)
101 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Preparation of highly functionalized alkylzinc halides from alkyl bromides using Mg, ZnCl2 and LiCl.
Chemical communications (Cambridge, England), 46(23), 4082-4084 (2010-04-10)
Alkyl bromides bearing various functional groups are smoothly converted to functionalized alkylzinc reagents by the direct insertion of magnesium in the presence of LiCl and ZnCl(2).
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service