Skip to Content
Merck
All Photos(1)

Documents

24008

Sigma-Aldrich

2-Chlorobutyric acid

≥97.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CHClCOOH
CAS Number:
Molecular Weight:
122.55
Beilstein:
1720944
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

≥97.0% (GC)

refractive index

n20/D 1.439

bp

90-92 °C/12 mmHg (lit.)

density

1.190 g/mL at 20 °C (lit.)

SMILES string

CCC(Cl)C(O)=O

InChI

1S/C4H7ClO2/c1-2-3(5)4(6)7/h3H,2H2,1H3,(H,6,7)

InChI key

RVBUZBPJAGZHSQ-UHFFFAOYSA-N

Application

2-Chlorobutyric acid was used in the synthesis of 2-hydroxy-5-hexenyl 2-chlorobutyrate ester.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

García et al.
Biochemical engineering journal, 5(3), 185-190 (2000-06-01)
An esterification process was developed for the direct synthesis of 2-hydroxy-5-hexenyl 2-chlorobutyrate ester from 2-chlorobutyric acid by using the epoxide 1,2-epoxy-5-hexene and Mucor miehei immobilised lipase as the biocatalyst in a batch reactor. The effect of temperature, catalyst concentration, and
Khatereh Bahrpaima et al.
Biomolecules, 9(8) (2019-08-23)
In this work, 1-carboxypropyled (1-CPRLS) and 5-carboxypentyled lignosulfonates (5-CPELS) were synthesized using 2-chlorobutanoic acid and 6-chlorohexanoic acid as carboxylate group donors via SN1 and SN2 mechanisms, respectively. 1-Carboxypropyl and 5-carboxypentyl lignosulfonates with the charge densities of -3.45 and -2.94 meq

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service