Skip to Content
Merck
All Photos(1)

Key Documents

237582

Sigma-Aldrich

Methyl 2-phenylsulfinylacetate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5S(O)CH2CO2CH3
CAS Number:
Molecular Weight:
198.24
Beilstein:
1953383
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

bp

130-131 °C/0.5 mmHg (lit.)

mp

52-54 °C (lit.)

SMILES string

COC(=O)CS(=O)c1ccccc1

InChI

1S/C9H10O3S/c1-12-9(10)7-13(11)8-5-3-2-4-6-8/h2-6H,7H2,1H3

InChI key

JPPXZUDQIXZLIL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Anion of methyl 2-phenylsulfinylacetate undergoes facile alkylative elimination under solid-liquid phase-transfer conditions to afford ring-substituted cinnamates[1]. Methyl 2-phenylsulfinylacetate undergoes condensation reactions with aldehydes to form vinylic sulfoxides[2].

Application

Anion of methyl 2-phenylsulfinylacetate has been employed in the synthesis of α,β-unsaturated esters[3].

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Journal of the Chemical Society. Perkin Transactions 1, 2683-2683 (1991)
New Synthetic reactions. Alkylative elimination.
Trost BM, et al.
Journal of the American Chemical Society, 96(22), 7165-7167 (1974)
Phase-Transfer Catalyzed Alkylative Elimination of Methyl 2-Phenylsulfinylacetate. A One-Pot Synthesis of Substituted Cinnamates.
Chong-ying X, et al.
Synthetic Communications, 17(15), 1839-1843 (1987)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service