Skip to Content
Merck
All Photos(2)

Key Documents

236128

Sigma-Aldrich

Cobalt(II) fluoride

powder

Synonym(s):

Cobaltous fluoride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CoF2
CAS Number:
Molecular Weight:
96.93
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
powder

grade

for analytical purposes

form

powder

concentration

37.8-40.6% (fluorine (F) basis (with lead acetate, gravimetric))

density

4.43 g/mL at 25 °C

application(s)

battery manufacturing

SMILES string

F[Co]F

InChI

1S/Co.2FH/h;2*1H/q+2;;/p-2

InChI key

YCYBZKSMUPTWEE-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ali Talimian et al.
Materials (Basel, Switzerland), 13(1) (2019-12-28)
Highly dense magnesium aluminate spinel bodies are usually fabricated using pressure-assisted methods, such as spark plasma sintering (SPS), in the presence of lithium fluoride as a sintering aid. The present work investigates whether the addition of transition metal fluorides promotes
Ru-Pan Wang et al.
The journal of physical chemistry. C, Nanomaterials and interfaces, 121(45), 24919-24928 (2017-11-25)
We show that with 2p3d resonant inelastic X-ray scattering (RIXS) we can accurately determine the charge-transfer parameters of CoF

Articles

Lithium-ion batteries' characteristics make them popular for electricity storage due to portability, rechargeability, and low cost.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service