Skip to Content
Merck
All Photos(1)

Key Documents

231231

Sigma-Aldrich

2-Methoxybenzonitrile

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H4CN
CAS Number:
Molecular Weight:
133.15
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

refractive index

n20/D 1.5465 (lit.)

bp

135 °C/12 mmHg (lit.)

density

1.093 g/mL at 25 °C (lit.)

functional group

nitrile

SMILES string

COc1ccccc1C#N

InChI

1S/C8H7NO/c1-10-8-5-3-2-4-7(8)6-9/h2-5H,1H3

InChI key

FSTPMFASNVISBU-UHFFFAOYSA-N

General description

Surface-enhanced Raman spectra study of 2-methoxybenzonitrile has been reported.

Application

2-Methoxybenzonitrile was used in the synthesis of 5-(4′-methyl [1,1′-biphenyl]-2-yl)-1H-tetrazole.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guillermo Diaz Fleming et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 71(3), 1074-1079 (2008-04-29)
The SERS modelling of o-, m-, and p-methoxybenzonitrile has been performed following the same methodology that in Part I. Optimized structure obtained from DFT calculations in a B3LYP-LANL2DZ level of calculation shows different tilted positions for the isomers under study.
[Catalysts for demethylation of methoxybenzonitrile in liquid-phase (author's transl)].
H Kashiwagi et al.
Yakugaku zasshi : Journal of the Pharmaceutical Society of Japan, 100(6), 668-671 (1980-06-01)
Efficient synthesis of 5-(4'-methyl [1, 1'-biphenyl]-2-yl)-1H-tetrazole.
Russell RK and Murray WV.
The Journal of Organic Chemistry, 58(18), 5023-5024 (1993)
Leandro Val Sayson et al.
Psychopharmacology, 236(7), 2201-2210 (2019-03-21)
Depressive syndrome or depression is a debilitating brain disorder affecting numerous people worldwide. Although readily available, current antidepressants have low remission rates and late onset times. Recently, N-methyl-D-aspartate (NMDA) receptor antagonists, like ketamine and methoxetamine (MXE), were found to elicit

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service