Skip to Content
Merck
All Photos(1)

Key Documents

227579

Sigma-Aldrich

4-Bromobutyl acetate

98%

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
R$677.00
50 G
R$1,805.00

R$677.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
R$677.00
50 G
R$1,805.00

About This Item

Linear Formula:
CH3CO2(CH2)4Br
CAS Number:
Molecular Weight:
195.05
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

R$677.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.46 (lit.)

bp

92-93 °C/12 mmHg (lit.)

density

1.348 g/mL at 25 °C (lit.)

functional group

bromo
ester

SMILES string

CC(=O)OCCCCBr

InChI

1S/C6H11BrO2/c1-6(8)9-5-3-2-4-7/h2-5H2,1H3

InChI key

UOABIRUEGSGTSA-UHFFFAOYSA-N

Application

4-Bromobutyl acetate was used in the synthesis of 9-alkylguanine derivative[1].

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

228.2 °F - closed cup

Flash Point(C)

109.00 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kjellberg J, et al.
Tetrahedron Letters, 27(7), 877-880 (1986)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service