Skip to Content
Merck
All Photos(3)

Key Documents

226548

Sigma-Aldrich

Silver perchlorate hydrate

99%

Synonym(s):

Perchloric acid silver hydrate, Silver perchlorate monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
AgClO4 · xH2O
CAS Number:
Molecular Weight:
207.32 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
form:
crystalline

Assay

99%

form

crystalline

reaction suitability

reagent type: oxidant

mp

485 °C (lit.)

storage temp.

2-8°C

SMILES string

O.[Ag+].[O-]Cl(=O)(=O)=O

InChI

1S/Ag.ClHO4.H2O/c;2-1(3,4)5;/h;(H,2,3,4,5);1H2/q+1;;/p-1

InChI key

IXFNJWPLHOWEPT-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Used to prepare silver 3,3′-dicyanodiphenylacetylene coordination networks for study of the effect of ligands on network structure toward the goal of engineering novel materials.

Pictograms

Flame over circleCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Ox. Sol. 2 - Skin Corr. 1B

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Keith A. Hirsch et al.
Inorganic chemistry, 36(14), 2960-2968 (1997-07-02)
Coordination networks of 3,3'-dicyanodiphenylacetylene (3,3'-DCPA, 1) with silver(I) salts characterized by single-crystal X-ray analysis are described. Network topology is found to depend on both the counterion and solvent employed during crystallization. The conformation adopted by the ligand varies between planar

Articles

Plasmonic nanoparticles have unique optical properties that can be tailored to suit a variety of applications in the biotechnology1–8 and electronics9–16 industries.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service