Skip to Content
Merck
All Photos(1)

Key Documents

226513

Sigma-Aldrich

B-Bromo-9-BBN solution

1.0 M in methylene chloride

Synonym(s):

9-Bromo-9-borabicyclo[3.3.1]nonane solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H14BBr
CAS Number:
Molecular Weight:
200.91
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

concentration

1.0 M in methylene chloride

density

1.315 g/mL at 25 °C

SMILES string

BrB1C2CCCC1CCC2

InChI

1S/C8H14BBr/c10-9-7-3-1-4-8(9)6-2-5-7/h7-8H,1-6H2/t7-,8+

InChI key

FYZDPIIMSZAKGP-OCAPTIKFSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service