Skip to Content
Merck
All Photos(1)

Key Documents

225428

Sigma-Aldrich

6-Chloro-2,4-dinitroaniline

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H2(NO2)2NH2
CAS Number:
Molecular Weight:
217.57
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

powder

mp

157-159 °C (lit.)

functional group

chloro

SMILES string

Nc1c(Cl)cc(cc1[N+]([O-])=O)[N+]([O-])=O

InChI

1S/C6H4ClN3O4/c7-4-1-3(9(11)12)2-5(6(4)8)10(13)14/h1-2H,8H2

InChI key

LHRIICYSGQGXSX-UHFFFAOYSA-N

General description

6-chloro-2,4-dinitroaniline exists in three polymorphic forms which can be separated on the basis of their mechanical properties.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Chronic 2 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Malla Reddy et al.
Chemical communications (Cambridge, England), (19)(19), 2439-2441 (2005-05-12)
Mechanical shearing of crystals of a polymorph with a layered structure may be used to separate them from harder crystals of a visually indistinguishable polymorph that resist such shearing.
Molecular crystals: pinching polymorphs.
Joel Bernstein
Nature materials, 4(6), 427-428 (2005-06-02)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service