Skip to Content
Merck
All Photos(1)

Documents

224251

Sigma-Aldrich

2-Propylpentanoic acid

98%

Synonym(s):

Valproic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2)2CHCO2H
CAS Number:
Molecular Weight:
144.21
EC Number:
MDL number:
UNSPSC Code:
12352103

Assay

98%

refractive index

n20/D 1.425 (lit.)

bp

220 °C (lit.)

density

0.9 g/mL at 25 °C (lit.)

SMILES string

CCCC(C(O)=O)CCC

InChI

1S/C8H16O2/c1-3-5-7(6-4-2)8(9)10/h7H,3-6H2,1-2H3,(H,9,10)

InChI key

NIJJYAXOARWZEE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Anticonvulsant that also has efficacy as a mood stabilizer in bipolar disorder

replaced by

Product No.
Description
Pricing

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service