Skip to Content
Merck
All Photos(1)

Key Documents

222127

Sigma-Aldrich

4-(Hexyloxy)phenol

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)5OC6H4OH
CAS Number:
Molecular Weight:
194.27
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

99%

form

solid

mp

45-47 °C (lit.)

SMILES string

CCCCCCOc1ccc(O)cc1

InChI

1S/C12H18O2/c1-2-3-4-5-10-14-12-8-6-11(13)7-9-12/h6-9,13H,2-5,10H2,1H3

InChI key

XIIIHRLCKLSYNH-UHFFFAOYSA-N

General description

4-(Hexyloxy)phenol is a lead anti-melanoma agent against B16-F0 melanoma cells with minimal metabolism by rat liver P450 microsomal preparation.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service