Skip to Content
Merck
All Photos(2)

Key Documents

221147

Sigma-Aldrich

Aminomalononitrile p-toluenesulfonate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CH(CN)2 · CH3C6H4SO3H
CAS Number:
Molecular Weight:
253.28
Beilstein:
4050747
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

98%

mp

174 °C (dec.) (lit.)

functional group

amine
nitrile
tosylate

SMILES string

NC(C#N)C#N.Cc1ccc(cc1)S(O)(=O)=O

InChI

1S/C7H8O3S.C3H3N3/c1-6-2-4-7(5-3-6)11(8,9)10;4-1-3(6)2-5/h2-5H,1H3,(H,8,9,10);3H,6H2

InChI key

MEUWQVWJLLBVQI-UHFFFAOYSA-N

Application

Aminomalononitrile p-toluenesulfonate was used in the syntheiss of 1-substituted 5-amino-4-cyano-2-hydroxyimidazoles[1].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kosaku Hirota et al.
Organic & biomolecular chemistry, 1(8), 1354-1365 (2003-08-22)
An efficient and general method for the synthesis of 2,9-disubstituted 8-hydroxyadenines, which are expected to have various biological activities, was realized. 5-Amino-4-cyano-2-hydroxyimidazoles(1) were prepared from aminomalononitrile and isocyanates as key intermediates. The condensation of 1a with amidines, imidates, guanidine, urea
Aleksandra Sochacka-Ćwikła et al.
Molecules (Basel, Switzerland), 25(15) (2020-08-08)
The synthesis of a series of novel 7-aminooxazolo[5,4-d]pyrimidines 5, transformations during their synthesis and their physicochemical characteristics have been described. Complete detailed spectral analysis of the intermediates 2-4, the N'-cyanooxazolylacetamidine by-products 7 and final compounds 5 has been carried out
Rou Jun Toh et al.
Langmuir : the ACS journal of surfaces and colloids, 35(30), 9896-9903 (2019-07-10)
In the last few years, the development of versatile coating chemistries has become a hot topic in surface science after the discovery that catecholamines can lead to conformal coatings upon oxidation from aqueous solutions. Recently, it was found that aminomalononitrile

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service