Skip to Content
Merck
All Photos(1)

Key Documents

220841

Sigma-Aldrich

cis-1,5-Dimethylbicyclo[3.3.0]octane-3,7-dione

≥98%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
R$587.00

R$587.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
R$587.00

About This Item

Empirical Formula (Hill Notation):
C10H14O2
CAS Number:
Molecular Weight:
166.22
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

R$587.00


Check Cart for Availability

Request a Bulk Order

Assay

≥98%

form

solid

mp

228-230 °C (lit.)

functional group

ketone

SMILES string

C[C@]12CC(=O)C[C@@]1(C)CC(=O)C2

InChI

1S/C10H14O2/c1-9-3-7(11)5-10(9,2)6-8(12)4-9/h3-6H2,1-2H3/t9-,10+

InChI key

KGZWVKASYOPUET-AOOOYVTPSA-N

General description

Olefination of cis-1,5-dimethylbicyclo[3.3.0]octane-3,7-dione with (carbethoxyethylidene)triphenylphospharane under controlled microwave heating has been investigated[1]. It reacts with carbon disulfide, strong base and an alkylating agent in dipolar aprotic solvents to yield racemic mixtures of 2-dialkylthio-methylene compounds[2].

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

DITHIOCARBOXYLIERUNGSREAKTIONEN VON cis-BICYCLO [3.3. 0] OCTAN-3, 7-DIONEN. MOLEKUL-UND KRISTALLSTRUKTUR VON cis-2, 6-BIS (DIMETHYLTHIO-METHYLEN)-BICYCLO [3.3. 0] OCTAN-3, 7-DION.
Dolling W, et al.
Phosphorus, Sulfur, and Silicon and the Related Elements, 113(1-4), 263-274 (1996)
Microwave-assisted regioselective olefinations of cyclic mono-and di-ketones with a stabilized phosphorus ylide.
Wu J, et al.
Tetrahedron, 62(19), 4643-4650 (2006)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service