Skip to Content
Merck
All Photos(1)

Key Documents

218944

Sigma-Aldrich

Diethylstilbestrol, mixture of cis and trans

97%

Synonym(s):

DES

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H20O2
CAS Number:
Molecular Weight:
268.35
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

170-172 °C (lit.)

SMILES string

CC\C(c1ccc(O)cc1)=C(\CC)c2ccc(O)cc2

InChI

1S/C18H20O2/c1-3-17(13-5-9-15(19)10-6-13)18(4-2)14-7-11-16(20)12-8-14/h5-12,19-20H,3-4H2,1-2H3/b18-17+

InChI key

RGLYKWWBQGJZGM-ISLYRVAYSA-N

Gene Information

General description

Mechanisms of the isomerization processes of molecular and anionic diethylstilbestrol (DES) has been reported. Estrogen activity of cis- and trans-diethylstilbestrol has been evaluated. Diethylstilbestrol is a carcinogenic synthetic estrogen.

Application

Diethylstilbestrol was used as internal standard for the analysis of drug residues in urine of rats and calves by on-line HPLC with ultraviolet and continuous-flow fast atom bombardmentmass spectrometry detectors.

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Diethylstilbestrol cis-trans isomerization and estrogen activity of diethylstilbestrol isomers.
V W Winkler et al.
Steroids, 17(2), 197-207 (1971-02-01)
E Davoli et al.
Analytical chemistry, 65(19), 2679-2685 (1993-10-01)
An automatic system for the on-line extraction and analysis of diethylstilbestrol in the urine of rats and calves is described. Extraction was done by injecting samples directly into an immunoaffinity column containing antidiethylstilbestrol antibodies bound to a Sepharose matrix, and
Osamu Takenouchi et al.
Bioconjugate chemistry, 27(11), 2689-2694 (2016-10-04)
Estrogens regulate different physiological systems with wide ranges of concentrations. The rapid analysis of estrogens is crucially important for drug discovery and medical diagnosis, but quantitation of nanomolar estrogens in live cells persists as an important challenge. We herein describe
Shoichi Kuramitsu et al.
JACC. Cardiovascular interventions, 8(9), 1180-1188 (2015-07-27)
This study sought to assess the incidence and clinical impact of stent fracture (SF) after the PROMUS Element platinum-chromium everolimus-eluting stent (PtCr-EES). SF remains an unresolved, clinically relevant issue, even in the newer-generation drug-eluting stent era. From March 2012 to
S T Jan et al.
Chemical research in toxicology, 11(5), 408-411 (1998-06-20)
Diethylstilbestrol (DES) and hexestrol (HES) are carcinogenic synthetic estrogens. The major metabolites of these compounds are their catechol derivatives, 3'-OH-DES and 3'-OH-HES. Oxidation of these metabolites leads to the electrophilic quinones, which are presumably involved in the tumor-initiating process. A

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service