212350
Chlorazol fast pink BK
Dye content 30 %
Synonym(s):
Benzo Fast Pink 2BL, Direct Red 75, Sirius Rose BB
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C33H22N8Na4O15S4
CAS Number:
Molecular Weight:
990.79
Colour Index Number:
25380
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
Recommended Products
composition
Dye content, 30%
λmax
522 nm
SMILES string
[Na+].[Na+].[Na+].[Na+].Nc1ccc2cc(cc(O)c2c1N=Nc3ccc(NC(=O)Nc4ccc(N=Nc5c(N)ccc6cc(cc(O)c56)S([O-])(=O)=O)c(c4)S([O-])(=O)=O)cc3S([O-])(=O)=O)S([O-])(=O)=O
Looking for similar products? Visit Product Comparison Guide
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
N S Doherty
Clinical and experimental immunology, 43(2), 260-269 (1981-02-01)
Chlorazole fast pink 2BL inhibited the classical complement pathway in rat serum both in vivo and in vitro. The in vitro potency of chlorazole fast pink against the alternative pathway could not be assessed accurately but it was clearly less
W J Fuggle et al.
Journal of clinical pathology, 37(3), 267-271 (1984-03-01)
Eosinophil polymorphonuclear leucocytes (polymorphs) were counted in 45 specimens from patients with Hodgkin's disease and five specimens from patients with reactive follicular hyperplasia. The use of chlorazol fast pink BK, a little known stain for eosinophil polymorphs, combined with image
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service