Skip to Content
Merck
All Photos(1)

Key Documents

210757

Sigma-Aldrich

Brilliant Crocein MOO

Dye content 70 %

Synonym(s):

7-Hydroxy-8-(4-phenylazophenylazo)-1,3-naphthalenedisulfonic acid disodium salt, Acid Red 73, Brilliant crocein scarlet MOO, Crocein Scarlet MOO

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H14N4Na2O7S2
CAS Number:
Molecular Weight:
556.48
Colour Index Number:
27290
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

composition

Dye content, 70%

λmax

510 nm

SMILES string

[Na+].[Na+].Oc1ccc2cc(cc(c2c1\N=N\c3ccc(cc3)\N=N\c4ccccc4)S([O-])(=O)=O)S([O-])(=O)=O

InChI

1S/C22H16N4O7S2.2Na/c27-19-11-6-14-12-18(34(28,29)30)13-20(35(31,32)33)21(14)22(19)26-25-17-9-7-16(8-10-17)24-23-15-4-2-1-3-5-15;;/h1-13,27H,(H,28,29,30)(H,31,32,33);;/q;2*+1/p-2/b24-23+,26-25+;;

InChI key

PEAGNRWWSMMRPZ-RVTJCSDESA-L

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fenglian Fu et al.
Chemosphere, 66(1), 1-7 (2006-07-11)
Three solid wastes, copper N,N'-bis(dithiocarboxy)piperazine ([CuBDP](n)), copper diethyldithiocarbamate (Cu(DDTC)(2)) and copper dimethyldithiocarbamate (Cu(DMTC)(2)), were prepared and tested as adsorbents to remove Acid Red 73 from wastewater. It was found that the three precipitates all could effectively adsorb the dye but
Chensi Shen et al.
Journal of hazardous materials, 177(1-3), 560-566 (2010-01-09)
A new efficient catalyst, CoTSPc@chitosan, was developed by immobilizing water soluble cobalt (II) tetrasulfophthalocyanine onto adsorbent chitosan microspheres covalently for the heterogeneous catalytic oxidation of C. I. Acid Red 73 with H(2)O(2). The result indicated that the COD removal and
The azo dye solvent yellow 3 produces depigmentation.
A K Bajaj et al.
Contact dermatitis, 42(4), 237-238 (2000-04-06)
Yuezhong Wen et al.
Journal of hazardous materials, 201-202, 162-169 (2011-12-16)
There is a need to explore effective and green approaches to enhancing the ability to use chitosan for contaminant removal for practical implementation of this technology. In the present study, glow discharge plasma (GDP), which has thus far been studied
C Gayathri et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 68(3), 578-582 (2007-02-28)
Organic dyes are very attractive optical materials for photonics and biophotonic applications. To quantitatively characterize their third-order nonlinear coefficients we have performed the experiments on two different azo dyes using a diode-pumped Nd:YAG laser at 532 nm. The |chi(3)| is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service