Skip to Content
Merck
All Photos(1)

Key Documents

209759

Sigma-Aldrich

2-Methylcyclopropanecarboxylic acid

98%

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
R$996.00

R$996.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
R$996.00

About This Item

Linear Formula:
CH3C3H4CO2H
CAS Number:
Molecular Weight:
100.12
Beilstein:
2323873
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

R$996.00


Check Cart for Availability

Request a Bulk Order

Assay

98%

form

liquid

refractive index

n20/D 1.439 (lit.)

bp

190-191 °C/745 mmHg (lit.)

density

1.027 g/mL at 25 °C (lit.)

functional group

carboxylic acid

SMILES string

CC1CC1C(O)=O

InChI

1S/C5H8O2/c1-3-2-4(3)5(6)7/h3-4H,2H2,1H3,(H,6,7)

InChI key

AYEGPMGNMOIHDL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

cis-2-methylcyclopropanecarboxylic acid is a component of curacin A[1].

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Efficient asymmetric synthesis of cis-2-methylcyclopropanecarboxylic acid.
Onoda T, et al.
Tetrahedron, 52(42), 13327-13338 (1996)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service