Skip to Content
Merck
All Photos(1)

Key Documents

209562

Sigma-Aldrich

2,4-Dinitrophenylacetic acid

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(O2N)2C6H3CH2CO2H
CAS Number:
Molecular Weight:
226.14
Beilstein:
1990798
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

mp

169-175 °C (lit.)

SMILES string

OC(=O)Cc1ccc(cc1[N+]([O-])=O)[N+]([O-])=O

InChI

1S/C8H6N2O6/c11-8(12)3-5-1-2-6(9(13)14)4-7(5)10(15)16/h1-2,4H,3H2,(H,11,12)

InChI key

KCNISYPADDTFDO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Coupling of 2,4-dinitrophenylacetic acid with diazonium salts has been reported.

Application

2,4-Dinitrophenylacetic acid was used in the preparation of 2-hydroxy-4-nitrohenzonitrile, required for the synthesis of parabactin azide (catecholamide siderophore photoaffinity label).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The coupling of diazonium salts with aliphatic carbon atoms.
Parmerter SM.
Org. React. (1959)
Guofeng Zhang et al.
Biochemistry, 41(45), 13370-13377 (2002-11-06)
Phosphonoacetaldehyde hydrolase (phosphonatase) from Bacillus cereus catalyzes hydrolytic P-C bond cleavage of phosphonoacetaldehyde (Pald) via a Schiff base intermediate formed with Lys53. A single turnover requires binding of Pald to the active site of the core domain, closure of the
A simple rapid mixing device.
J F Eccleston et al.
Analytical biochemistry, 106(1), 73-77 (1980-07-15)
Synthesis of a parabactin photoaffinity label.
Bergeron RJ, et al.
The Journal of Organic Chemistry, 52(1), 144-149 (1987)
A G Lowe et al.
Analytical biochemistry, 144(2), 385-389 (1985-02-01)
A quenched-flow apparatus is described and applied to measurements of the hydrolysis of 2,4-dinitrophenyl acetate by sodium hydroxide and the entry of D-[U-14C]glucose into human red blood cells at 37 degrees C. Glucose influx into red cells was a saturable

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service