Skip to Content
Merck
All Photos(1)

Key Documents

19997

Sigma-Aldrich

tert-Butyl hydroperoxide solution

packed in FEP bottles, ~5.5 M in decane (over molecular sieve 4Å)

Synonym(s):

1,1-Dimethylethyl hydroperoxide, 2-Hydroperoxy-2-methylpropane, TBHP

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3COOH
CAS Number:
Molecular Weight:
90.12
Beilstein:
1098280
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

liquid

quality

packed in FEP bottles

reaction suitability

reagent type: oxidant

concentration

52.9-66.8% (iodometric)
~5.5 M in decane (over molecular sieve 4Å)

refractive index

n20/D 1.403

density

0.83 g/mL at 20 °C

storage temp.

2-8°C

SMILES string

CC(C)(C)OO

InChI

1S/C4H10O2/c1-4(2,3)6-5/h5H,1-3H3

InChI key

CIHOLLKRGTVIJN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Used in oxidative desulfurization of fuels[1]and as an oxidant for the epoxidation of styrene.[2]

Other Notes

Safe and stable oxidant for the Sharpless epoxidation[3][4]

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 2 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 3 - Muta. 2 - Org. Perox. F - Skin Corr. 1C - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.2 - Organic peroxides and self-reacting hazardous materials

WGK

WGK 3

Flash Point(F)

109.4 °F - closed cup

Flash Point(C)

43 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Oxidative desulfurization of fuel oil. Part I. Oxidation of dibenzothiphenes using tert-butyl hydroperoxide
Wang D,et al.
Applied Catalysis A: General, 253, 91-99 (2003)
Controlled synthesis of a- and c-Fe2O3 nanoparticles via thermolysis of PVA gels and studies on a-Fe2O3 catalyzed styrene epoxidation.
Bepari RA,et al.
Journal of Saudi Chemical Society (2014)
K. Akashi et al.
The Journal of Organic Chemistry, 43, 2063-2063 (1978)
Qicai Xue et al.
Chemical communications (Cambridge, England), 49(35), 3700-3702 (2013-03-29)
A new synthetic approach toward direct C-N bond formation through sp(3) C-H activation has been developed under metal-free conditions. Both primary and secondary benzylic C-H substrates could react smoothly with various amines to give only mono-amination products with good to
Younghwa Kim et al.
Food chemistry, 137(1-4), 136-141 (2012-12-04)
Oligomeric and polymeric procyanidins have been reported to possess different antioxidant capacities. However, the intracellular antioxidant mechanisms of oligomeric and polymeric procyanidins are still poorly understood. In this study, we evaluated the cytoprotective effects of the oligomeric procyanidin fraction (OPF)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service