Skip to Content
Merck
All Photos(1)

Key Documents

193437

Sigma-Aldrich

2-Methylcyclohexylamine, mixture of cis and trans

98%

Synonym(s):

1-Amino-2-methylcyclohexane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H10NH2
CAS Number:
Molecular Weight:
113.20
Beilstein:
2689073
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.4565 (lit.)

bp

149-150 °C (lit.)

density

0.856 g/mL at 25 °C (lit.)

SMILES string

CC1CCCCC1N

InChI

1S/C7H15N/c1-6-4-2-3-5-7(6)8/h6-7H,2-5,8H2,1H3

InChI key

FEUISMYEFPANSS-UHFFFAOYSA-N

General description

Mechanism of hydrodenitrogenation of 2-methylcyclohexylamine has been studied over sulfided NiMo/Al2O3. 2-Methylcyclohexylamine is the intermediate formed during hydrodenitrogenation of toluidine.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

102.2 °F - closed cup

Flash Point(C)

39 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mechanism of the C−N-bond breaking in the hydrodenitrogenation of methylcyclohexylamine over sulfided NiMo/γ-Al2O3.
Rota F and Prins R.
J. Mol. Catal. A: Chem., 162(1), 367-374 (2000)
Mechanisms of the hydrodenitrogenation of alkylamines with secondary and tertiary a-carbon atoms on sulfided NiMo/Al2O3.
Zhao Y and Prins R.
J. Catal., 222(2), 532-544 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service