Skip to Content
Merck
All Photos(1)

Key Documents

190616

Sigma-Aldrich

2-Hydroxy-3-nitropyridine

98%

Synonym(s):

3-Nitro-2-pyridinol, 3-Nitro-2-pyridone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H4N2O3
CAS Number:
Molecular Weight:
140.10
Beilstein:
124474
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

212 °C (dec.) (lit.)

SMILES string

Oc1ncccc1[N+]([O-])=O

InChI

1S/C5H4N2O3/c8-5-4(7(9)10)2-1-3-6-5/h1-3H,(H,6,8)

InChI key

BOAFCICMVMFLIT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Hydroxy-3-nitropyridine reacts with phosphorus pentachloride and phosphoryl chloride to yield 2-chloro-3-nitropyridine.

Application

2-Hydroxy-3-nitropyridine was used in the synthesis of:
  • 3-nitro-2-pyridyl β-D-galactoside
  • 3-nitro-2-pyridyl N-acetyl-β-D-glucosaminide
  • 5-amino-6-chloro-3-iodopyridine

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nitropyridyl glycosides: new glycosyl donors for enzymatic transglycosylation.
Yasukochi T, et al.
Tetrahedron Letters, 40(36), 6585-6589 (1999)
Nanjing Zhang et al.
Bioorganic & medicinal chemistry letters, 13(3), 525-527 (2003-02-05)
Racemic 5-azidoepibatidine [(+/-)-1] was synthesized via 5-aminoepibatidine as a candidate photoaffinity ligand with exceptionally high affinity at the mammalian neuronal nicotinic receptors (K(i) values of 0.027 nM for alpha 4 beta 2 and 9.7 nM for alpha 7) and excellent
85. Nucleophilic displacement reactions in aromatic systems. Part I. Kinetics of the reactions of chloronitropyridines with aromatic amines and with pyridine.
Bishop RR, et al.
Journal of the Chemical Society, 437-446 (1952)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service