Skip to Content
Merck
All Photos(1)

Key Documents

189790

Sigma-Aldrich

2,3,4-Trimethoxybenzoic acid

≥98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)3C6H2CO2H
CAS Number:
Molecular Weight:
212.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

≥98%

form

solid

mp

99-102 °C (lit.)

functional group

carboxylic acid

SMILES string

COc1ccc(C(O)=O)c(OC)c1OC

InChI

1S/C10H12O5/c1-13-7-5-4-6(10(11)12)8(14-2)9(7)15-3/h4-5H,1-3H3,(H,11,12)

InChI key

HZNQSWJZTWOTKM-UHFFFAOYSA-N

Application

2,3,4-Trimethoxybenzoic acid was used in the synthesis of tropoloisoquinoline alkaloid pareitropone[1]. It was also used in the synthesis of isomeric tris(pyrogallol) derivatives[2] and naphthoic acid[3].

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and characterization of a series of vanadium-tunichrome B 1 analogs. Crystal structure of a tris (catecholamide) complex of vanadium.
Bulls AR, et al.
Journal of the American Chemical Society, 112(7), 2627-2632 (1990)
Hung-Sheng Soung et al.
Neurotoxicity research, 34(3), 375-387 (2018-04-10)
Reserpine (RES)-induced orofacial dyskinesia (OD) has been used as an animal model for human tardive dyskinesia (TD) for decades, due to its strong pathophysiological association with striatal oxidative stress and neural cytoarchitecture alteration. L-Theanine (LT), one of the major amino
Ken S Feldman et al.
Journal of the American Chemical Society, 124(39), 11600-11601 (2002-09-26)
The synthesis of the tropoloisoquinoline alkaloid pareitropone has been accomplished in 14 steps from 2,3,4-trimethoxybenzoic acid. The key transformations include the generation of an alkylidenecarbene intermediate through intramolecular addition of a tosylamide anion to an alkynyliodonium salt, and the cycloaddition
An oxazoline based approach to (S)-Gossypol.
Meyers AI and Willemsen JJ.
Tetrahedron, 54(35), 10493-10511 (1998)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service