Skip to Content
Merck
All Photos(1)

Key Documents

188565

Phthalimidoacetaldehyde dimethyl acetal

98%

Synonym(s):

N-(2,2-Dimethoxyethyl)phthalimide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H13NO4
CAS Number:
Molecular Weight:
235.24
PubChem Substance ID:

Assay

98%

InChI

1S/C12H13NO4/c1-16-10(17-2)7-13-11(14)8-5-3-4-6-9(8)12(13)15/h3-6,10H,7H2,1-2H3

InChI key

HDSDOJMUZFFCFU-UHFFFAOYSA-N


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service