Skip to Content
Merck
All Photos(1)

Key Documents

185981

Sigma-Aldrich

4-Nitrophenyl phenyl sulfide

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4SC6H5
CAS Number:
Molecular Weight:
231.27
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

form

solid

mp

52-55 °C (lit.)

SMILES string

[O-][N+](=O)c1ccc(Sc2ccccc2)cc1

InChI

1S/C12H9NO2S/c14-13(15)10-6-8-12(9-7-10)16-11-4-2-1-3-5-11/h1-9H

InChI key

RJCBYBQJVXVVKB-UHFFFAOYSA-N

General description

4-Nitrophenyl phenyl sulfide participates in copper-catalysed S-arylation of thiophenol derivatives with aryl halides[1].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T R Juneja et al.
Mutation research, 263(1), 13-19 (1991-05-01)
4-Nitrodiphenyl thioether (4-NO2TE) and its various potential metabolites 4-nitroso- (4-NOTE), 4-hydroxylamino- (4-NHOHTE), 4-hydroxyacetylamino- [4-N(OH)ACTE], 4-acetoxy-acetylamino- [4-N(OAc)-ACTE], 4-amino- (4-ATE) and 4-acetamido- (4-AATE) were tested for their mutagenicity toward Salmonella typhimurium TA98 in the presence and absence of S9 activation system. 4-NO2TE
Mónica Carril et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 13(18), 5100-5105 (2007-03-28)
A novel protocol for the copper-catalysed S-arylation of thiophenol derivatives with aryl halides leading to diaryl sulfides is reported. The reactions were catalysed by a combination of a copper salt and a 1,2-diamine derivative (acting both as the ligand and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service