Skip to Content
Merck
All Photos(1)

Documents

183741

Sigma-Aldrich

(+)-p-Menth-1-en-9-ol,mixture of isomers

(R,R)-isomer ~67%, (R,S)-isomer ~33%, 97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18O
CAS Number:
Molecular Weight:
154.25
EC Number:
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:

Assay

97%

optical activity

[α]20/D +99°, c = 4.2 in toluene

refractive index

n20/D 1.486 (lit.)

bp

115-116 °C/10 mmHg (lit.)

density

0.941 g/mL at 25 °C (lit.)

SMILES string

CC(CO)C1CCC(C)=CC1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

217.4 °F - closed cup

Flash Point(C)

103 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service