Skip to Content
Merck
All Photos(3)

Key Documents

183334

Sigma-Aldrich

Ammonium benzoate

98%

Synonym(s):

Benzoic acid ammonium salt

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
R$298.00
1 KG
R$476.00

R$298.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 G
R$298.00
1 KG
R$476.00

About This Item

Linear Formula:
C6H5COONH4
CAS Number:
Molecular Weight:
139.15
Beilstein:
3634518
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

R$298.00


Check Cart for Availability

Request a Bulk Order

Assay

98%

form

solid

mp

192-198 °C (dec.) (lit.)

functional group

carboxylic acid
phenyl

SMILES string

N.OC(=O)c1ccccc1

InChI

1S/C7H6O2.H3N/c8-7(9)6-4-2-1-3-5-6;/h1-5H,(H,8,9);1H3

InChI key

VWSRWGFGAAKTQG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ligang Deng et al.
Molecules (Basel, Switzerland), 25(3) (2020-01-26)
The experiment developed the ultra-high-performance liquid chromatography-tandem mass spectrometry (UPLC/MS/MS) method for testing emamectin benzoate, and studied the metabolism of emamectin benzoate in rice plants and rice-growing environments via application of this testing method. The dissipation curve of emamectin benzoate
A system of qualitative analysis for the common metals in presence of phosphates, using ammonium benzoate.
G L AUSTIN
The Analyst, 72(859), Unknown-Unknown (1947-10-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service