Skip to Content
Merck
All Photos(1)

Key Documents

183040

Sigma-Aldrich

2-(Benzyloxy)phenol

96%

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
R$478.00

R$478.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
R$478.00

About This Item

Linear Formula:
C6H5CH2OC6H4OH
CAS Number:
Molecular Weight:
200.23
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

R$478.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

96%

form

liquid

impurities

<3% catechol

refractive index

n20/D 1.591 (lit.)

density

1.143 g/mL at 25 °C (lit.)

SMILES string

Oc1ccccc1OCc2ccccc2

InChI

1S/C13H12O2/c14-12-8-4-5-9-13(12)15-10-11-6-2-1-3-7-11/h1-9,14H,10H2

InChI key

CCZCXFHJMKINPE-UHFFFAOYSA-N

Application

2-(Benzyloxy)phenol was used in the synthesis of 2-(benzyloxy)hydroquinone[1].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

No data available

Flash Point(C)

No data available

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Woro Triarsi Sulistyaningdyah et al.
Applied microbiology and biotechnology, 67(4), 556-562 (2004-11-19)
Cytochrome P450 BM-3 from Bacillus megaterium is a fatty acid hydroxylase exhibiting selectivity for long-chain substrates (12-20 carbons). Replacement of Phe87 in P450 BM-3 by Val (F87V) greatly increased its activity towards a variety of aromatic and phenolic compounds. The

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service