Skip to Content
Merck
All Photos(1)

Key Documents

180556

Sigma-Aldrich

trans,trans-2,4-Nonadienal

≥85%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3CH=CHCH=CHCHO
CAS Number:
Molecular Weight:
138.21
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor density

>1 (vs air)

Assay

≥85%

refractive index

n20/D 1.5207 (lit.)

bp

97-98 °C/10 mmHg (lit.)

density

0.862 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

[H]C(=O)\C([H])=C([H])\C([H])=C(/[H])CCCC

InChI

1S/C9H14O/c1-2-3-4-5-6-7-8-9-10/h5-9H,2-4H2,1H3/b6-5+,8-7+

InChI key

ZHHYXNZJDGDGPJ-BSWSSELBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

trans,trans-2,4-Nonadienal forms adduct in the presence of tetrahydrofuran (THF).

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

186.8 °F - closed cup

Flash Point(C)

86 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ana Paula M Loureiro et al.
Chemical research in toxicology, 18(2), 290-299 (2005-02-22)
The reaction of 2'-deoxyguanosine with the alpha,beta-unsaturated aldehydes trans-2-octenal, trans-2-nonenal, trans-2-decenal, trans,trans-2,4-nonadienal, and trans,trans-2,4-decadienal in THF gives rise to three novel adducts: 3-(2'-deoxy-beta-D-erythro-pentafuranosyl)-7-[3-hydroxy-1-(3-(2'-deoxy-beta-D-erythro-pentafuranosyl)-3,5-dihydro-imidazo[1,2-a]purin-9-one-7-yl)-propyl]-3,5-dihydro-imidazo[1,2-a]purin-9-one (A7) and 3-(2'-deoxy-beta-D-erythro-pentafuranosyl)-7-(tetrahydrofuran-2-yl)-3,5-dihydro-imidazo[1,2-a]purin-9-one (A8 and A9), which are not observed in the absence of THF. These adducts
Virginia C Resconi et al.
Food chemistry, 242, 288-300 (2017-10-19)
This study aims to assess the changes in beef aroma over time when steaks from pre-aged knuckles are stored in retail display under high oxygen conditions for 15 or 22days in vacuum conditions. Odorous volatile compounds were analysed by solid-phase

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service