Skip to Content
Merck
All Photos(1)

Key Documents

178446

Sigma-Aldrich

(2,2-Dichlorocyclopropyl)benzene

97%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
R$632.00

R$632.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
R$632.00

About This Item

Linear Formula:
C6H5C3H3Cl2
CAS Number:
Molecular Weight:
187.07
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

R$632.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

refractive index

n20/D 1.551 (lit.)

bp

103 °C/10 mmHg (lit.)

density

1.208 g/mL at 25 °C (lit.)

functional group

chloro
phenyl

SMILES string

ClC1(Cl)CC1c2ccccc2

InChI

1S/C9H8Cl2/c10-9(11)6-8(9)7-4-2-1-3-5-7/h1-5,8H,6H2

InChI key

WLWFQGXZIDYWQF-UHFFFAOYSA-N

Application

(2,2-Dichlorocyclopropyl)benzene may be used in the synthesis of p-(2,2-dichlorocyclopropyl)aniline.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

219.2 °F - closed cup

Flash Point(C)

104.00 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service