Skip to Content
Merck
All Photos(1)

Key Documents

177121

Sigma-Aldrich

3-Quinolinecarboxaldehyde

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H7NO
CAS Number:
Molecular Weight:
157.17
Beilstein:
113269
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

68-71 °C (lit.)

functional group

aldehyde

SMILES string

[H]C(=O)c1cnc2ccccc2c1

InChI

1S/C10H7NO/c12-7-8-5-9-3-1-2-4-10(9)11-6-8/h1-7H

InChI key

RYGIHSLRMNXWCN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The structure and vibrational spectra of 3-quinolinecarboxaldehyde has been studied[1].

Application

3-Quinolinecarboxaldehyde was used in the synthesis of 1,4-addition products[2].

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

M Kumru et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 90, 28-34 (2012-02-07)
Combined experimental and theoretical studies have been performed on the structure and vibrational spectra (IR and Raman spectra including far region) of 3-quinolinecarboxaldehyde. Hartree-Fock (HF) and density functional theory (DFT) B3LYP calculations have been employed with the 6-311++G(d,p) basis set
Ferles M and Kocian O.
Collection of Czechoslovak Chemical Communications, 46(6), 1518-1522 (1981)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service