Skip to Content
Merck
All Photos(1)

Key Documents

165670

Sigma-Aldrich

Methyl 2,4-dimethoxybenzoate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)2C6H3CO2CH3
CAS Number:
Molecular Weight:
196.20
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.548 (lit.)

bp

294-296 °C (lit.)

density

1.194 g/mL at 25 °C (lit.)

SMILES string

COC(=O)c1ccc(OC)cc1OC

InChI

1S/C10H12O4/c1-12-7-4-5-8(10(11)14-3)9(6-7)13-2/h4-6H,1-3H3

InChI key

IHIJFZWLGPEYPJ-UHFFFAOYSA-N

General description

Methyl 2,4-dimethoxybenzoate has highest antifeedant index and are the best candidates for practical applications in order to protect planted seedlings in the field.

Application

Methyl 2,4-dimethoxybenzoate was used in the synthesis of 1,5-diaryl-1,3,5-pentanetriones.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

New Strong Base Synthesis of Symmetrical 1, 5-Diaryl-1, 3, 5-pentanetriones from Acetone and Benzoate Esters.
Knight JD, et al.
Synthetic Communications, 38(14), 2465-2482 (2008)
C Rikard Unelius et al.
Journal of chemical ecology, 32(10), 2191-2203 (2006-10-03)
Aromatic organic compounds found in the feces of the pine weevil, Hylobius abietis (L.) (Coleoptera: Curculionidae), have been shown to deter feeding behavior in this species, which is a serious pest of planted conifer seedlings in Europe. We evaluated 55

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service