Skip to Content
Merck
All Photos(1)

Key Documents

164984

Sigma-Aldrich

8-Hydroxyquinoline

≥99%

Synonym(s):

8-Quinolinol, 8-Hydroxyquinoline, 8-Oxychinolin, Oxine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H7NO
CAS Number:
Molecular Weight:
145.16
Beilstein:
114512
EC Number:
MDL number:
UNSPSC Code:
12352100

Assay

≥99%

mp

72.5-74.0 °C

antibiotic activity spectrum

fungi

Mode of action

DNA synthesis | interferes
enzyme | inhibits

SMILES string

Oc1cccc2cccnc12

InChI

1S/C9H7NO/c11-8-5-1-3-7-4-2-6-10-9(7)8/h1-6,11H

InChI key

MCJGNVYPOGVAJF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: quinolone

Application

Metal-chelating agent

replaced by

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Repr. 1B - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

María Isabel Fernández-Bachiller et al.
Journal of medicinal chemistry, 53(13), 4927-4937 (2010-06-16)
Tacrine and PBT2 (an 8-hydroxyquinoline derivative) are well-known drugs that inhibit cholinesterases and decrease beta-amyloid (Abeta) levels by complexation of redox-active metals, respectively. In this work, novel tacrine-8-hydroxyquinoline hybrids have been designed, synthesized, and evaluated as potential multifunctional drugs for
Mahmud Tareq Hassan Khan et al.
Journal of medicinal chemistry, 52(1), 48-61 (2008-12-17)
In the present work, 22 compounds of the U.S. NCI compound library (size 273K) were identified as putative thermolysin binders by structure based virtual screening with the ICM software (ICM-VLS). In vitro competitive binding assays confirmed that 12 were thermolysin
David M Rubush et al.
Journal of the American Chemical Society, 134(33), 13554-13557 (2012-08-09)
The desymmetrization of p-peroxyquinols using a Brønsted acid-catalyzed acetalization/oxa-Michael cascade was achieved in high yields and selectivities for a variety of aliphatic and aryl aldehydes. Mechanistic studies suggest that the reaction proceeds through a dynamic kinetic resolution of the peroxy
Bhimashankar Mitkari et al.
Experimental neurology, 239, 158-162 (2012-10-13)
Cell therapies from various sources have been under intense research in stroke. Efficient homing of the cells to the injured brain without complications is necessary to realize the therapeutic potential of cell therapy. Intra-arterial (IA) infusion of cells bypasses the
Erik H J G Aarntzen et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 19(6), 1525-1533 (2013-02-06)
Anticancer dendritic cell (DC) vaccines require the DCs to relocate to lymph nodes (LN) to trigger immune responses. However, these migration rates are typically very poor. Improving the targeting of ex vivo generated DCs to LNs might increase vaccine efficacy

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service