Skip to Content
Merck
All Photos(3)

Key Documents

161721

Sigma-Aldrich

3′,5′-Dimethoxyacetophenone

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)2C6H3COCH3
CAS Number:
Molecular Weight:
180.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

refractive index

n20/D 1.543 (lit.)

bp

290-291 °C (lit.)

mp

33-34 °C (lit.)

functional group

ketone

SMILES string

COc1cc(OC)cc(c1)C(C)=O

InChI

1S/C10H12O3/c1-7(11)8-4-9(12-2)6-10(5-8)13-3/h4-6H,1-3H3

InChI key

YJKHOUIVWKQRSL-UHFFFAOYSA-N

Application

3′,5′-Dimethoxyacetophenone was used as starting reagent in the synthesis of 5,7-dimethoxy-3-methylindazole. It was used in the synthesis of (R)-(+)-1-(3,5-dimethoxyphenyl)ethyl acetate-ether-180.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stereochemistry of photosolvolysis of a chiral, 18O-labeled 1-arylethyl acetate.
Jaeger DA and Angelos GH
Tetrahedron Letters, 22(9), 803-806 (1981)
5, 7-Dimethoxy-3-Methylindazole from 3, 5-Dimethoxyacetophenone.
Boudreault N and Leblanc Y.
Organic Syntheses, 241-241 null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service