Skip to Content
Merck
All Photos(1)

Key Documents

161268

Sigma-Aldrich

4-Phenyl-3-butyn-2-one

96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C≡CCOCH3
CAS Number:
Molecular Weight:
144.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

form

liquid

refractive index

n20/D 1.574 (lit.)

bp

75-76 °C/0.8 mmHg (lit.)

density

0.99 g/mL at 25 °C (lit.)

functional group

ketone

storage temp.

2-8°C

SMILES string

CC(=O)C#Cc1ccccc1

InChI

1S/C10H8O/c1-9(11)7-8-10-5-3-2-4-6-10/h2-6H,1H3

InChI key

UPEUQDJSUFHFQP-UHFFFAOYSA-N

General description

4-Phenyl-3-butyn-2-one is an α,β-ketoalkyne. Reaction of 4-phenyl-3-butyn-2-one with bromine chloride and iodine monochloride in CH2Cl2, CH2Cl2/pyridine and MeOH are studied. Reduction of 4-phenyl-3-butyn-2-one in THF solution has been reported.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

203.0 °F - closed cup

Flash Point(C)

95 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Improved procedure for the asymmetric reduction of prochiral ketones by B-3-pinanyl-9-borabicyclo [3.3. 1] nonane.
Brown HC and Pai GG.
The Journal of Organic Chemistry, 47(8), 1606-1608 (1982)
Victor L Heasley et al.
The Journal of organic chemistry, 67(7), 2183-2187 (2002-04-02)
The reactions of 3-butyn-2-one (1), 3-hexyn-2-one (2), and 4-phenyl-3-butyn-2-one (3) with bromine chloride (BrCl) and iodine monochloride (ICl) in CH(2)Cl(2), CH(2)Cl(2)/pyridine, and MeOH are described. The data show that the major products in CH(2)Cl(2) are (Z)-AM (anti-Markovnikov) regioisomers. With the
Shigeyuki Kitamura et al.
Drug metabolism and disposition: the biological fate of chemicals, 30(4), 414-420 (2002-03-20)
The reduction of the triple bond and carbonyl group of an alpha,beta-ketoalkyne, 4-phenyl-3-butyn-2-one (PBYO), by rat liver microsomes and cytosol was investigated. The triple-bond-reduced product trans-4-phenyl-3-buten-2-one (PBO) and the carbonyl-reduced product 4-phenyl-3-butyn-2-ol (PBYOL) were formed when PBYO was incubated with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service