161195
2-Methoxy-5-nitroaniline
98%
Synonym(s):
5-Nitro-o-anisidine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Assay
98%
mp
117-119 °C (lit.)
SMILES string
COc1ccc(cc1N)[N+]([O-])=O
InChI
1S/C7H8N2O3/c1-12-7-3-2-5(9(10)11)4-6(7)8/h2-4H,8H2,1H3
InChI key
NIPDVSLAMPAWTP-UHFFFAOYSA-N
General description
2-Methoxy-5-nitroaniline is an aromatic metabolite of 2,4-dinitroanisole.
Application
2-Methoxy-5-nitroaniline was used in the synthesis of 5-(9-acridinylamino)-p-anisidines via reaction with 9-anilinoacridines. It was also used in the synthesis of disazo disperse dyes containing nitro and methoxy groups, used for the dyeing of polyester fibre.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
246.2 °F - closed cup
Flash Point(C)
119 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
5-Nitro-ortho-anisidine.
IARC monographs on the evaluation of the carcinogenic risk of chemicals to humans, 27, 133-139 (1982-04-01)
Biometrics, 40(1), 27-40 (1984-03-01)
An index of carcinogenic potency for chemicals tested in chronic animal experiments is described. By analogy with the well-known 'lethal dose 50' (LD50) of quantal bioassay, a 'tumorigenic dose 50' (TD50) may be defined (in the absence both of tumors
Disperse dyes derived from 2-methoxy-5-nitroaniline.
Orient. J. Chem., 25(4), 863-863 (2009)
Journal of hazardous materials, 262, 281-287 (2013-09-18)
2,4-Dinitroanisole (DNAN) is an insensitive munitions compound considered to replace conventional explosives such as 2,4,6-trinitrotoluene (TNT). DNAN undergoes facile microbial reduction to 2-methoxy-5-nitroaniline (MENA) and 2,4-diaminoanisole (DAAN). This study investigated the inhibitory effect of DNAN, MENA, and DAAN toward various
Physical chemistry chemical physics : PCCP, 17(34), 22072-22078 (2015-08-04)
Reductive degradation of three typical multi-substituted nitroaromatic pollutants by zero valent iron was comprehensively compared in terms of performance, kinetics, toxicity and mechanisms in this study. The results showed that 0.5 mM 2,4-dinitrochlorobenzene (DNCB), 2,4-dinitroanisole (DNAN) and 2,4-dinitrophenol (DNP) could
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service