Skip to Content
Merck
All Photos(1)

Documents

160652

Sigma-Aldrich

1-Bromo-2,6-dichlorobenzene

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H3Cl2
CAS Number:
Molecular Weight:
225.90
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

powder

bp

242 °C/765 mmHg (lit.)

mp

61-64 °C (lit.)

SMILES string

Clc1cccc(Cl)c1Br

InChI

1S/C6H3BrCl2/c7-6-4(8)2-1-3-5(6)9/h1-3H

InChI key

UWOIDOQAQPUVOH-UHFFFAOYSA-N

General description

Extent of debromination of 1-bromo-2,6-dichlorobenzene in several aprotic solvents has been evaluated.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reductive debromination of 1-methyl-2, 4, 5-tribromoimidazole mediated by dry tetramethylammonium fluoride in aprotic solvents.
Bastos MM, et al.
Journal of the Brazilian Chemical Society, 12(3), 417-421 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service