Skip to Content
Merck
All Photos(1)

Key Documents

158852

Sigma-Aldrich

7-Chloro-4-hydrazinoquinoline

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H8ClN3
CAS Number:
Molecular Weight:
193.63
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

219-225 °C (dec.) (lit.)

SMILES string

NNc1ccnc2cc(Cl)ccc12

InChI

1S/C9H8ClN3/c10-6-1-2-7-8(13-11)3-4-12-9(7)5-6/h1-5H,11H2,(H,12,13)

InChI key

TWJRTPKEHPDDPG-UHFFFAOYSA-N

Application

7-Chloro-4-hydrazinoquinoline was used in the preparation of new Schiff base hydrazone ligand via reaction with o-hydroxybenzaldehyde. It was also used in the preparation of N′-(E)-heteroaromatic-isonicotinohydrazide derivatives and the heteroaromatic 7-chloro-4-quinolinylhydrazone derivatives.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mostafa El-Behery et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 66(1), 28-36 (2006-11-23)
A new hydrazone ligand, HL, was prepared by the reaction of 7-chloro-4-hydrazinoquinoline with o-hydroxybenzaldehyde. The ligand behaves as monoprotic bidentate. This was accounted for as the ligand contains a phenolic group and its hydrogen atom is reluctant to be replaced
Marcelle de L Ferreira et al.
TheScientificWorldJournal, 10, 1347-1355 (2010-07-14)
Two series of N'-(E)-heteroaromatic-isonicotinohydrazide derivatives (3a-f and 4a-b) and 1-(7-chloroquinolin-4-yl)-2-[(heteroaromatic)methylene]hydrazone derivatives (5a-f and 6a-b) have been synthesized and evaluated for their in vitro antibacterial activity against Mycobacterium tuberculosis H37Rv. Several compounds were noncytotoxic and exhibited significant minimum inhibitory concentration (MIC)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service