157171
2-Nitrodiphenylamine
98%
Synonym(s):
2-Nitro-N-phenylaniline
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
vapor density
10.7 (vs air)
Assay
98%
mp
74-76 °C (lit.)
SMILES string
[O-][N+](=O)c1ccccc1Nc2ccccc2
InChI
1S/C12H10N2O2/c15-14(16)12-9-5-4-8-11(12)13-10-6-2-1-3-7-10/h1-9,13H
InChI key
RUKISNQKOIKZGT-UHFFFAOYSA-N
General description
2-Nitrodiphenylamine is a double-base (DB) rocket propellant stabilizer and its microencapsulation potentially improves the shelf life of DB rocket propellants. It is one of the component of Otto Fuel II, a propellant in torpedoes. Solubilization of 2-nitrodiphenylamine, a hydrophobic polar dye molecule, in aqueous solutions of polystyrene(310)-b-poly(acrylic acid) micelles was investigated.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Journal of microencapsulation, 29(6), 569-575 (2012-08-24)
We demonstrate the microencapsulation of a double-base (DB) rocket propellant stabiliser, 2-nitrodiphenylamine (2-NDPA), that can potentially increase the shelf life of DB rocket propellants. Poly(lactide-co-glycolide) (PLG) microspheres loaded with 2-NDPA were prepared using the oil-in-water emulsion technique. The microsphere size
Journal of the American Chemical Society, 125(39), 11993-12000 (2003-09-25)
We investigate the solubilization of 2-nitrodiphenylamine, a hydrophobic but polar dye molecule, in aqueous solutions of polystyrene(310)-b-poly(acrylic acid)(47) micelles. The solubilization capacity of the micelles, which consist of a polystyrene core and poly(acrylic acid) corona, and the micelle-water partition coefficient
Anaerobe, 4(2), 95-102 (2006-08-05)
Otto Fuel II, a propellant in torpedoes, is composed of 76% 1,2 propanediol dinitrate (PGDN), 22.5% di-n-butyl sebacate, and 1.5% 2-nitrodiphenylamine (NDPA), and is largely recalcitrant to aerobic microbial degradation. Anaerobic microbial degradation of Otto Fuel II was tested by
Analytical and bioanalytical chemistry, 410(14), 3349-3360 (2018-04-05)
Monitoring specific chemical properties is the key to chemical process control. Today, mainly optical online methods are applied, which require time- and cost-intensive calibration effort. NMR spectroscopy, with its advantage being a direct comparison method without need for calibration, has
Forensic science international, 301, 129-136 (2019-06-04)
The detection and analysis of gunshot residues expelled during the discharge of the weapon play a potential role in criminal investigations. In this work, organic gunshot residues produced by using automatic rifle Kalashnikov caliber: 7.62×39mm and semi-automatic pistol Makarov caliber:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service