Skip to Content
Merck
All Photos(2)

Key Documents

155349

Sigma-Aldrich

Thallium(I) cyclopentadienide

97%

Synonym(s):

5-Cyclopentadienyl)thallium, Cyclopentadienylthallium, NSC 114719, Thallous cyclopentadienide

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
R$1,279.00

R$1,279.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
R$1,279.00

About This Item

Empirical Formula (Hill Notation):
C5H5Tl
CAS Number:
Molecular Weight:
269.48
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

R$1,279.00


Check Cart for Availability

Request a Bulk Order

Assay

97%

reaction suitability

core: thallium
reagent type: catalyst
reaction type: Olefin Metathesis

mp

300 °C (lit.)

storage temp.

2-8°C

SMILES string

[Tl].[CH]1[CH][CH][CH][CH]1

InChI

1S/C5H5.Tl/c1-2-4-5-3-1;/h1-5H;

InChI key

RKLJDPOVAWGBSD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reagent in preparation of
  • η5-Cyclopentadienylpalladium(II) complexes as catalysts for vinyl addition polymerization and copolymerization reactions
  • Versatile carbon ligands via oxidative cyclization
  • Isomeric dinuclear dicyclopentadienyl Ti chloride complexes containing bridging cyclopentadienylsiloxo ligands with Me Li and Lewis acids
  • Bromide complexes of the triple-decker type
  • Naphthylazo cyclopalladates via regiospecific naphthyl carbon-hydrogen bond activation

Reactant fro:
  • Selective reduction of the U3+ metallocene amidinate

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Chronic 2 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service