Skip to Content
Merck
All Photos(1)

Documents

154571

Sigma-Aldrich

1-Methoxynaphthalene

≥98%

Synonym(s):

Methyl 1-naphthyl ether, NSC 5530

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H7OCH3
CAS Number:
Molecular Weight:
158.20
Beilstein:
774884
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98%

form

liquid

refractive index

n20/D 1.621 (lit.)

bp

135-137 °C/12 mmHg (lit.)

density

1.09 g/mL at 25 °C (lit.)

SMILES string

COc1cccc2ccccc12

InChI

1S/C11H10O/c1-12-11-8-4-6-9-5-2-3-7-10(9)11/h2-8H,1H3

InChI key

NQMUGNMMFTYOHK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1-Methoxynaphthalene was used to study the peroxygenase activity of CcP. It was also used to synthesize prenyl naphthalen-ols.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

233.6 °F - closed cup

Flash Point(C)

112 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

James E Erman et al.
BMC biochemistry, 14, 19-19 (2013-07-31)
The cytochrome P450s are monooxygenases that insert oxygen functionalities into a wide variety of organic substrates with high selectivity. There is interest in developing efficient catalysts based on the "peroxide shunt" pathway in the cytochrome P450s, which uses H2O2 in
Kazutoshi Shindo et al.
Bioscience, biotechnology, and biochemistry, 75(3), 505-510 (2011-03-11)
We performed combinational bioconversion of substituted naphthalenes with PhnA1A2A3A4 (an aromatic dihydroxylating dioxygenase from marine bacterium Cycloclasticus sp. strain A5) and prenyltransferase NphB (geranyltransferase from Streptomyces sp. strain CL190) or SCO7190 (dimethylallyltransferase from Streptomyces coelicolor A3(2)) to produce prenyl naphthalen-ols.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service