Skip to Content
Merck
All Photos(1)

Key Documents

153729

Sigma-Aldrich

2-Ethoxybenzaldehyde

≥97%

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
R$537.00

R$537.00


Estimated to ship onMay 21, 2025


Request a Bulk Order

Select a Size

Change View
25 G
R$537.00

About This Item

Linear Formula:
C2H5OC6H4CHO
CAS Number:
Molecular Weight:
150.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

R$537.00


Estimated to ship onMay 21, 2025


Request a Bulk Order

Assay

≥97%

refractive index

n20/D 1.543 (lit.)

bp

136-138 °C/24 mmHg (lit.)

density

1.074 g/mL at 25 °C (lit.)

functional group

aldehyde

SMILES string

[H]C(=O)c1ccccc1OCC

InChI

1S/C9H10O2/c1-2-11-9-6-4-3-5-8(9)7-10/h3-7H,2H2,1H3

InChI key

DUVJMSPTZMCSTQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Ethoxybenzaldehyde was used to study the 1,3-dipolar cycloaddition reactions[1].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

197.6 °F - closed cup

Flash Point(C)

92 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rate accelerations of 1, 3-dipolar cycloaddition reactions in ionic liquids.
Dubreuil JF and Bazureau JP.
Tetrahedron Letters, 41(38), 7351-7355 (2000)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service