Skip to Content
Merck
All Photos(2)

Documents

15268

Sigma-Aldrich

N-Boc-diethanolamine

≥98.0% (GC)

Synonym(s):

tert-Butyl N,N-bis(2-hydroxyethyl)carbamate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H19NO4
CAS Number:
Molecular Weight:
205.25
Beilstein:
4386562
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0% (GC)

reaction suitability

reagent type: cross-linking reagent

refractive index

n20/D 1.460

density

1.085 g/mL at 20 °C (lit.)

SMILES string

CC(C)(C)OC(=O)N(CCO)CCO

InChI

1S/C9H19NO4/c1-9(2,3)14-8(13)10(4-6-11)5-7-12/h11-12H,4-7H2,1-3H3

InChI key

KMUNFRBJXIEULW-UHFFFAOYSA-N

Other Notes

Building block for the synthesis of cationic lipids, nucleosides, etc; monomer for synthesis of poly(urethanes)

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

I. Solodin et al.
Synlett, 617-617 (1996)
S C Bergmeier et al.
Nucleosides & nucleotides, 18(2), 227-238 (1999-03-06)
We have prepared a series of novel aza-acyclonucleosides as potential antiviral agents. These compounds were prepared from diethanolamine and the desired purine or pyrimidine base via a Mitsunobu coupling. No antiviral activity was observed against either HSV-1 or HCMV.
J.M. Orban et al.
Journal of Polymer Science Part A: Polymer Chemistry, 37, 3441-3441 (1999)
Cláudia D Raposo et al.
Polymers, 12(1) (2020-01-18)
Doxorubicin-loaded PLGA nanoparticles conjugated with a new galactose-based ligand for the specific recognition by human hepatoma cellular carcinoma cells (Hep G2) were successfully produced. The new targeting compound was selected using molecular docking combined with quantum chemical calculations for modelling

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service