Skip to Content
Merck
All Photos(1)

Key Documents

149446

Sigma-Aldrich

1-Pyrrolidino-1-cyclopentene

98%

Synonym(s):

1-(1-Cyclopenten-1-yl)pyrrolidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H15N
CAS Number:
Molecular Weight:
137.22
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

98%

form

liquid

refractive index

n20/D 1.5155 (lit.)

bp

100-110 °C/15 mmHg (lit.)

density

0.941 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

C1CCN(C1)C2=CCCC2

InChI

1S/C9H15N/c1-2-6-9(5-1)10-7-3-4-8-10/h5H,1-4,6-8H2

InChI key

KOFSFYBXUYHNJL-UHFFFAOYSA-N

Application

1-pyrrolidino-1-cyclopentene was used in the synthesis of halichlorine, pinnaic acid, and tauropinnaic acid[1].

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

116.6 °F - closed cup

Flash Point(C)

47 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hamish S Christie et al.
Proceedings of the National Academy of Sciences of the United States of America, 101(33), 12079-12084 (2004-08-10)
The related marine natural products halichlorine, pinnaic acid, and tauropinnaic acid have been synthesized. The described route provided access to all three compounds from a common, late-stage intermediate. The synthesis began with 1-pyrrolidino-1-cyclopentene from which an intermediate possessing the three

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service