14931
Boc-(S)-α-(4-fluorobenzyl)-Pro-OH
≥98.0% (HPLC)
Synonym(s):
(S)-1-Boc-2-(4-fluorobenzyl)-2-pyrrolidinecarboxylic acid, Boc-α-(4-fluorobenzyl)-D-proline
About This Item
Recommended Products
product name
Boc-(S)-α-(4-fluorobenzyl)-Pro-OH, ≥98.0% (HPLC)
Assay
≥98.0% (HPLC)
form
powder or crystals
color
white to almost white
application(s)
peptide synthesis
SMILES string
CC(C)(C)OC(=O)N1CCC[C@]1(Cc2ccc(F)cc2)C(O)=O
InChI
1S/C17H22FNO4/c1-16(2,3)23-15(22)19-10-4-9-17(19,14(20)21)11-12-5-7-13(18)8-6-12/h5-8H,4,9-11H2,1-3H3,(H,20,21)/t17-/m0/s1
InChI key
AWTXCFLBYUGCEW-KRWDZBQOSA-N
Signal Word
Warning
Hazard Statements
Hazard Classifications
Acute Tox. 4 Oral
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Proline analogues are promising candidates for tuning the biological, pharmaceutical, or physicochemical properties of naturally occuring, as well as de novo designed, linear, and, cyclic peptides.
Proline analogues are promising candidates for tuning the biological, pharmaceutical, or physicochemical properties of naturally occuring, as well as de novo designed, linear, and, cyclic peptides.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service